Please login to your online account to display your discounted pricing

GR 113808, Tocris

Potent, selective 5-HT4 antagonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material GR 113808
Assay Percent Range >98%
Molecular Formula C19H27N3O4S
Formula Weight 393.5
Synonym 1-methyl-1H-indole-3-carboxylic acid, [1-[2-[(methylsulfonyl)amino]ethyl]-4-piperidinyl]methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

132250

 
r & d systems
RDS132250
50mg Each for $924.23

132210

 
r & d systems
RDS132210
10mg Each for $225.77
Description & Specifications

Specifications

Formula Weight 393.5
Quantity 50mg
Assay Percent Range >98%
CAS 144625-51-4
Recommended Storage Store at Room Temperature
Chemical Name or Material GR 113808
Molecular Formula C19H27N3O4S
Synonym 1-methyl-1H-indole-3-carboxylic acid, [1-[2-[(methylsulfonyl)amino]ethyl]-4-piperidinyl]methyl ester
Safety and Handling

Store at Room Temperature