Please login to your online account to display your discounted pricing

GR 113808, Tocris


Potent, selective 5-HT4 antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS132210

 View more versions of this product

Catalog No. 132210

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 393.5
Quantity 10mg
Assay Percent Range >98%
Chemical Name or Material GR 113808
CAS 144625-51-4
Synonym 1-methyl-1H-indole-3-carboxylic acid, [1-[2-[(methylsulfonyl)amino]ethyl]-4-piperidinyl]methyl ester
Molecular Formula C19H27N3O4S
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material GR 113808
Assay Percent Range >98%
Molecular Formula C19H27N3O4S
Formula Weight 393.5
Synonym 1-methyl-1H-indole-3-carboxylic acid, [1-[2-[(methylsulfonyl)amino]ethyl]-4-piperidinyl]methyl ester
Accessories

Store at Room Temperature