Please login to your online account to display your discounted pricing

GR 103691, Tocris

Highly selective D3 antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material GR 103691
Assay Percent Range >98%
Molecular Formula C30H35N3O3
Formula Weight 485.63
Synonym 4'-Acetyl-N-[4-[4-(2-methoxyphenyl)-1-piperazinyl]butyl]-[1, 1'-biphenyl]-4-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

110910

 
r & d systems
110910
10mg Each for $201.36

110950

 
r & d systems
110950
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 485.63
Quantity 10mg
Chemical Name or Material GR 103691
CAS 162408-66-4
Synonym 4'-Acetyl-N-[4-[4-(2-methoxyphenyl)-1-piperazinyl]butyl]-[1, 1'-biphenyl]-4-carboxamide
Molecular Formula C30H35N3O3
Assay Percent Range >98%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature