missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ GoTaq™ G2 Hot Start Green Master Mixes DFS Item

GoTaq™ G2 Hot Start Master Mixes are ready-to-use mixes containing GoTaq™ G2 Hot Start Polymerase, buffer, dNTPs and optimized magnesium

Manufacturer:  Promega™ M7423

 View more versions of this product

Catalog No. PRM7423



Description

Description

  • Contains a gel loading dye to facilitate downstream gel analysis
  • Second generation of GoTaq products
  • Is bound to a proprietary antibody that blocks activity
  • Activity is restored during initial denaturation, allowing hot-start PCR
  • Robust reliable PCR for all your amplification needs
  • Simplify reaction setup and save time with a ready-to-use master mix
  • Prepare your reaction at room temperature, not on ice
  • Eliminate nonspecific amplification with hot-start enzyme
  • Use at no risk—backed by the Promega PCR Satisfaction Guarantee
Specifications

Specifications

1000 reactions
Green master mix
−30° to −10°C
Documents
Promotions

Promotions