missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega GoTaq™ G2 Colorless Master Mix
GoTaq(R) G2 Master Mixes are premixed ready-to-use solutions containing GoTaq(R) G2 DNA Polymerase, dNTPs, MgCl2 and reaction buffers at optimal concentrations for efficient amplification of a wide range of DNA templates by PCR.
Supplier: Promega M7833
Description
- All-in-One PCR Master Mix: Just Add Template and Primers
- High-Performance GoTaq(R) G2 DNA Polymerase in a Ready-to-Use Master Mix
- Simplifies reaction setup and saves time
- Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required
Specifications
−30° to −10°C | |
1000 reactions | |
Colorless master mix (M783B 1 x 25mL), Nuclease-free water (P119C 1 x 25mL) |
Colorless master mix | |
RT-PCR |