missing translation for 'onlineSavingsMsg'
Learn More

Gossypol-acetic acid

CAS: 12542-36-8 | C32H34O10 | 578.614 g/mol

$144.23 - $144.23

Chemical Identifiers

CAS 12542-36-8
Molecular Formula C32H34O10
Molecular Weight (g/mol) 578.614
MDL Number MFCD00058385
InChI Key NIOHNDKHQHVLKA-UHFFFAOYSA-N
Synonym gossypol-acetic acid, gossypol acetate, gossypol acetic acid, acetate gossypol, --gossypol acetic acid, s-gossypol acetic acid, at-101 acetic acid, gossypol-acetic acid complex
PubChem CID 227456
IUPAC Name acetic acid;7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
SMILES CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C.CC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63517MC
View Documents
Thermo Scientific Chemicals
J63517MC
100 mg
Each for $144.23
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12542-36-8
578.614
NIOHNDKHQHVLKA-UHFFFAOYSA-N
227456
CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C.CC(=O)O
C32H34O10
MFCD00058385
gossypol-acetic acid, gossypol acetate, gossypol acetic acid, acetate gossypol, --gossypol acetic acid, s-gossypol acetic acid, at-101 acetic acid, gossypol-acetic acid complex
acetic acid;7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde

Specifications

12542-36-8
C32H34O10
100 mg
Soluble in acetone or methanol at 5mg/ml
CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C.CC(=O)O
578.614
578.61
Gossypol-acetic acid
Yellow to Green
MFCD00058385
gossypol-acetic acid, gossypol acetate, gossypol acetic acid, acetate gossypol, --gossypol acetic acid, s-gossypol acetic acid, at-101 acetic acid, gossypol-acetic acid complex
NIOHNDKHQHVLKA-UHFFFAOYSA-N
acetic acid;7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
227456
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H351

RTECSNumber : MD7360000

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only