missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gossypol, 98+%

Lipophilic agent derived from cottonseed

$217.43 - $471.89

Chemical Identifiers

CAS 303-45-7
Molecular Formula C30H30O8
Molecular Weight (g/mol) 518.562
MDL Number MFCD00017352
InChI Key QBKSWRVVCFFDOT-UHFFFAOYSA-N
Synonym gossypol, --gossypol, +-gossypol, pogosin, tash 1, racemic-gossypol, r---gossypol, r-gossypol, +--gossypol, +/--gossypol
PubChem CID 3503
ChEBI CHEBI:28584
IUPAC Name 7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
SMILES CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63767MC View Documents Alfa Aesar™
J63767MC
100mg Each for $217.43
 
AAJ63767MD View Documents Alfa Aesar™
J63767MD
250mg Each for $471.89
 
Description

Description

Applications
Gossypol acts as an inhibitor for several dehydrogenase enzymes. Gossypol has also long been known to possess antimalarial properties. Gossypol provided reliable contraception. gossypol provokes infertility and causes spermatogenesis. Gossypol is used as an active antifertility agent in a variety of male animals, including mice, rats, hamsters, monkeys, rabbits, and bulls.

Solubility
Soluble in 100%ethanol (25 mg/ml), DMF (25 mg/ml), acetone, DMSO (25 mM), methanol (2 mg/ml), ether, chloroform, sodium carbonate, and dilute aqueous solutions of ammonia . Insoluble in water. Gossypol is a male antifertility agent with antispermatogenic activity and has been shown to contain antitumor, anitviral, and antioxidant properties.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

303-45-7
518.562
QBKSWRVVCFFDOT-UHFFFAOYSA-N
3503
7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
C30H30O8
MFCD00017352
gossypol, --gossypol, +-gossypol, pogosin, tash 1, racemic-gossypol, r---gossypol, r-gossypol, +--gossypol, +/--gossypol
CHEBI:28584
CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C

Specifications

Yellow to Brown
303-45-7
MFCD00017352
gossypol, --gossypol, +-gossypol, pogosin, tash 1, racemic-gossypol, r---gossypol, r-gossypol, +--gossypol, +/--gossypol
CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C
518.562
CHEBI:28584
Powder
Gossypol
100mg
C30H30O8
14,4528
QBKSWRVVCFFDOT-UHFFFAOYSA-N
7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
3503
518.56
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H341-H361-H302
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.

GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DU3100000

Recommended Storage : −30° to −10°C