Corning™ Sterile Polystyrene Spreader: L-Shaped
Designed for use in industrial microbiology labs such as food and beverage, environmental/water testing.
Manufacturer:  Corning™ ETAR01
 View more versions of this product
Catalog No. 07-201-928
Description
Description
  • Polystyrene (PS)
  • Packed in resealable bag
  • Curved end or 'T' end
  • Sterile
'
Specifications
Specifications
CE, EC N-1935/2004, ISO 11137- sterile SAL 10-3
5000/Cs.
L-shaped
14.8 cm
Polystyrene
Sterile
White
1000/Pk.
5.82 in.
Designed for use in industrial microbiology labs such as food and beverage, environmental/water testing
Spreader