missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ 1 mL Straw Pipet, Polypropylene, Sterile

Decrease cross contamination risks using disposable pipets

Manufacturer:  Corning™ EPI24B02

 View more versions of this product

Catalog No. 07-201-924



Description

Description

  • Product traceability on each bag
  • Sterile 10-3
  • Packaged individually in tamper evident single zip bags

Specifications

Specifications

Yes
6000/Cs.
Polypropylene
Straw Pipet
50/Bag
24 cm
Sterile
2.2 mL
SDS
Documents