missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Sterile Polyethylene Blender Bags
Universal bags for homogeneous suspension
Manufacturer: Corning™ BBAG03
Description
- Polyethylene (PE) bags
- 80 to 3500 mL
- With or without filter
- Traceability on each filter bag
- Sterile
Order Info
Use bag clip, Cat. No. 07-201-979, to close bags between sample preparation steps.
Specifications
With lateral filter | |
500/Pk. | |
400 mL | |
300 mm | |
Sterile |
1500/Cs. | |
25/Pk. | |
Natural | |
PE (Polyethylene) | |
190 mm |