missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Sterile Polyethylene Blender Bags

Universal bags for homogeneous suspension

Manufacturer:  Corning™ SM301

 View more versions of this product

Catalog No. 07-201-921



Description

Description

  • Polyethylene (PE) bags
  • 80 to 3500 mL
  • With or without filter
  • Traceability on each filter bag
  • Sterile

Order Info

Use bag clip, Cat. No. 07-201-979, to close bags between sample preparation steps.

Specifications

Specifications

Without filter
100/Pk.
3500 mL
510 mm
Sterile
500/Cs.
25/Pk.
Natural
PE (Polyethylene)
380 mm
SDS
Documents