missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Round-Bottom Petri Dish
Optimized for automation
Manufacturer: Corning™ BB6402
Description
- 65 x 14.5 mm (D x H)
- 0/3/4 vents
- Crystal Polystyrene (PS)
- Optimal flatness
- Grip lid
- Molded grid 10x10 mm
Specifications
14.5 mm | |
65 mm | |
Polystyrene | |
Gridded | |
20/Pk. | |
1 |
Round | |
Sterile | |
Cell Culture Petri Dish | |
720/Cs. | |
Yes |