Please login to your online account to display your discounted pricing
Gosselin™ Petri Dishes
Optimized for automation. Gosselin™ Petri Dishes are manufactured from crystal polystyrene.
Manufacturer: gosselin/corning inc  BP93D-01
 View all options for this product
Catalog No. 07-201-910
Description & Specifications
Diameter (Metric) 100mm
Shape Round
Vent 3 Vents
Height (Metric) 15mm
Compartments 1.0
Color Transparent
Packaging Double bagged
Sterile No
Material Polystyrene (PS)
  • Crystal polystyrene
  • Optimal flatness
  • Manufactured in class 100
  • Easy opening