Please login to your online account to display your discounted pricing

Gosselin™ Autoclavable Blender Bags


For autoclaving contaminated products safely

Manufacturer: corning incorporated  SA0905

 View more versions of this product

Catalog No. 07-201-964

  • / Case of 750 Each
 

Description & Specifications

Specifications

For Use With (Application) Clinical, dairy, environment, industries, microbiology
Includes Biohazard symbol
Thickness 40μm
Autoclavable Yes
Width (English) 11.81 in.
Height (English) 19.68 in.
Height (Metric) 50cm
Capacity (English) 2.37 gal.
Capacity (Metric) 9L
Material HDPE
Color Natural
Capacity 2.37 gal., 9L
Width (Metric) 30cm