Please login to your online account to display your discounted pricing

Gosselin™ Autoclavable Blender Bags


For autoclaving contaminated products safely

Manufacturer: corning incorporated  SA0305

 View more versions of this product

Catalog No. 07-201-962

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Color Natural
Thickness (Metric) 40μm
Capacity (English) 0.79 gal.
Width (Metric) 25.5cm
Material HDPE (High-density Polyethylene)
Includes Biohazard symbol
Width (English) 10.03 in.
Autoclavable Yes
Height (English) 15.74 in.
Capacity (Metric) 3L
Length (English) 0.79 gal.
For Use With (Application) Clinical, dairy, environment, industries, microbiology
Height (Metric) 40cm

  • Thickness: 40μm
  • Features wide range of volumes