Please login to your online account to display your discounted pricing

Gosselin™ Autoclavable Blender Bags


For autoclaving contaminated products safely

Manufacturer: corning incorporated  SA3303

 View more versions of this product

Catalog No. 07-201-970

  • / Case of 500 Each
 

Description & Specifications

Specifications

For Use With (Application) Clinical, dairy, environment, industries, microbiology
Height (English) 31.49 in.
Height (Metric) 80cm
Includes Biohazard symbol
Thickness (Metric) 40μm
Width (English) 19.68 in.
Autoclavable Yes
Capacity (English) 8.71 gal.
Capacity (Metric) 33L
Color Natural
Material HDPE (High-density Polyethylene)
Width (Metric) 50cm