Please login to your online account to display your discounted pricing

Gosselin™ Autoclavable Blender Bags


For autoclaving contaminated products safely

Manufacturer: corning incorporated  SA2105

 View more versions of this product

Catalog No. 07-201-968

  • / Case of 400 Each
 

Description & Specifications

Specifications

Autoclavable Yes
Width (English) 15.74 in.
For Use With (Application) Clinical, dairy, environment, industries, microbiology
Height (English) 30.7 in.
Height (Metric) 78cm
Includes Biohazard symbol
Thickness 40μm
Width (Metric) 40cm
Capacity (English) 5.54 gal.
Capacity (Metric) 21L
Material HDPE
Color Natural
Capacity 5.54 gal., 21L