missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Autoclavable Blender Bags
For autoclaving contaminated products safely
Manufacturer: Corning™ SA1403
Description
- Polypropylene (PP) resistant bags
- Thermal resistance 134°C
- Transparent
Specifications
23.62 in. | |
Autoclavable | |
9L | |
Clinical, dairy, environment, industries, microbiology | |
HDPE | |
15.74 in. |
60 cm | |
2.37 gal. | |
Natural | |
Biohazard symbol | |
40μm | |
40 cm |