Please login to your online account to display your discounted pricing

Golgin-97 Mouse anti-Human, eFluor 660, Clone: CDF4, eBioscience™

Mouse Monoclonal Antibody

$160.95 - $390.05

Specifications

Antigen Golgin-97
Clone CDF4
Host Species Murine
Gene Alias Golgin Subfamily A Member 1, Golgin A1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124535

 
life technologies
LS50976780
25μg Each for $160.95

501124536

 
life technologies
LS50976782
100μg Each for $390.05
Description & Specifications

Specifications

Antigen Golgin-97
Clone CDF4
Host Species Murine
Gene Alias Golgin Subfamily A Member 1, Golgin A1
Species Reactivity Human
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody CDF4 recognizes human Golgin-97. Golgin-97, a GRIP domain protein member that was first identified as a Golgi complex autoantigen associated with Sjogren’s syndrome. Golgin-97 is a peripheral membrane protein found on the trans face of the Golgi apparatus. The protein G family member, Arl1, mediates the binding of Golgin-97 to the Golgi membrane. Like all GRIP proteins, Golgin-97 functions in maintaining the integrity of the trans Golgi network. Golgin-97 has also been shown to function as a tethering molecule in the retrograde transport of proteins from the endosomes to the trans Golgi network. During poxvirus infection, Golgin-97 incorporates into virions and plays a role in viral replication and morphogenesis.