Please login to your online account to display your discounted pricing

Golgin-97 Mouse anti-Human, Clone: CDF4, eBioscience™

Mouse Monoclonal Antibody

$146.45 - $368.30

Specifications

Antigen Golgin-97
Clone CDF4
Host Species Murine
Gene Alias Golgin Subfamily A Member 1, Golgin A1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122845

 
life technologies
LS14976780
25μg Each for $146.45

501122846

 
life technologies
LS14976782
100μg Each for $368.30
Description & Specifications

Specifications

Antigen Golgin-97
Clone CDF4
Host Species Murine
Gene Alias Golgin Subfamily A Member 1, Golgin A1
Species Reactivity Human
Applications Immunocytochemistry
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody CDF4 recognizes human Golgin-97. Golgin-97, a GRIP domain protein member that was first identified as a Golgi complex autoantigen associated with Sjogren’s syndrome. Golgin-97 is a peripheral membrane protein found on the trans face of the Golgi apparatus. The protein G family member, Arl1, mediates the binding of Golgin-97 to the Golgi membrane. Like all GRIP proteins, Golgin-97 functions in maintaining the integrity of the trans Golgi network. Golgin-97 has also been shown to function as a tethering molecule in the retrograde transport of proteins from the endosomes to the trans Golgi network. During poxvirus infection, Golgin-97 incorporates into virions and plays a role in viral replication and morphogenesis.