Please login to your online account to display your discounted pricing

Golden Syrian Hamster anti-Ly-49C/I/F/H, Clone: 14B11, PE, Novus Biologicals™

$246.00 - $386.00

Specifications

Antigen Ly-49C/I/F/H
Clone 14B11
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is NK cell (Ly-49C/I/F/H).
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20019510

 
novus biologicals canada
NBP200195100UG
100μg Each for $246.00

NBP20019526

 
novus biologicals canada
NBP200195200UG
200μg Each for $386.00
Description & Specifications

Specifications

Antigen Ly-49C/I/F/H
Clone 14B11
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is NK cell (Ly-49C/I/F/H).
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Golden Syrian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry