Please login to your online account to display your discounted pricing

Golden Syrian Hamster anti-Ly-49C/I/F/H, Clone: 14B11, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200195100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20019510

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Primary or Secondary Primary
Immunogen The immunogen for this antibody is NK cell (Ly-49C/I/F/H).
Conjugate PE
Monoclonal or Polyclonal Monoclonal
Clone 14B11
Cross Reactivity Mouse
Isotype IgG
Concentration 0.2mg/mL
Purification Method Affinity Purified
Antigen Ly-49C/I/F/H
Quantity 100μg
Storage Requirements Store at 4°C. Do not freeze.
Host Species Golden Syrian Hamster
Format Affinity Purified
Regulatory Status RUO