Please login to your online account to display your discounted pricing

Golden Syrian Hamster anti-Ly-49C/I/F/H, Clone: 14B11, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200194500UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20019455

  • / Each
 

Description & Specifications

Specifications

Antigen Ly-49C/I/F/H
Host Species Golden Syrian Hamster
Isotype IgG
Purification Method Affinity Purified
Cross Reactivity Mouse
Format Affinity Purified
Applications Flow Cytometry
Immunogen The immunogen for this antibody is NK cell (Ly-49C/I/F/H).
Quantity 500μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 14B11
Conjugate FITC
Concentration 0.5mg/mL