Please login to your online account to display your discounted pricing

Golden Syrian Hamster anti-Ly-49C/I/F/H, Clone: 14B11, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200194100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20019410

  • / Each
 

Description & Specifications

Specifications

Host Species Golden Syrian Hamster
Applications Flow Cytometry
Immunogen The immunogen for this antibody is NK cell (Ly-49C/I/F/H).
Isotype IgG
Quantity 100μg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Clone 14B11
Cross Reactivity Mouse
Antigen Ly-49C/I/F/H
Format Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Concentration 0.5mg/mL
Conjugate FITC
Purification Method Affinity Purified
Primary or Secondary Primary