Please login to your online account to display your discounted pricing

Golden Syrian Hamster anti-Ly-49C/I/F/H, Clone: 14B11, Biotin, Novus Biologicals™

$192.00 - $493.00

Specifications

Antigen Ly-49C/I/F/H
Clone 14B11
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is NK cell (Ly-49C/I/F/H).
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20019610

 
novus biologicals canada
NBP200196100UG
100μg Each for $192.00

NBP20019655

 
novus biologicals canada
NBP200196500UG
500μg Each for $493.00
Description & Specifications

Specifications

Antigen Ly-49C/I/F/H
Clone 14B11
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is NK cell (Ly-49C/I/F/H).
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Golden Syrian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry