Please login to your online account to display your discounted pricing

Golden Syrian Hamster anti-Ly-49C/I/F/H, Clone: 14B11, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200196500UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20019655

  • / Each
 

Description & Specifications

Specifications

Antigen Ly-49C/I/F/H
Host Species Golden Syrian Hamster
Quantity 500μg
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Applications Flow Cytometry
Immunogen The immunogen for this antibody is NK cell (Ly-49C/I/F/H).
Isotype IgG
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Clone 14B11
Concentration 0.5mg/mL
Conjugate Biotin
Cross Reactivity Mouse
Purification Method Affinity Purified
Primary or Secondary Primary