Please login to your online account to display your discounted pricing

Golden Syrian Hamster anti-Ly-49C/I/F/H, Clone: 14B11, Biotin, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP200196100UG

 View all options for this product


For Research Use Only

Catalog No. NBP20019610


Description & Specifications


Antigen Ly-49C/I/F/H
Applications Flow Cytometry
Clone 14B11
Concentration 0.5mg/mL
Conjugate Biotin
Cross Reactivity Mouse
Format Affinity Purified
Host Species Golden Syrian Hamster
Immunogen The immunogen for this antibody is NK cell (Ly-49C/I/F/H).
Isotype IgG
Purification Method Affinity Purified
Quantity 100μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal