Please login to your online account to display your discounted pricing

Golden Syrian Hamster anti-CD28, Clone: 37.51, Biotin, Novus Biologicals™

$252.00 - $472.00

Specifications

Antigen CD28
Clone 37.51
Concentration 0.5mg/mL
Immunogen Mouse T cell lymphoma EL-4 cells
Gene ID (Entrez) 940
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20010210

 
novus biologicals canada
NBP200102100UG
100μg Each for $252.00

NBP20010255

 
novus biologicals canada
NBP200102500UG
500μg Each for $472.00
Description & Specifications

Specifications

Antigen CD28
Clone 37.51
Concentration 0.5mg/mL
Immunogen Mouse T cell lymphoma EL-4 cells
Gene ID (Entrez) 940
Formulation Protein G purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Golden Syrian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Biotin
Format Purified
Purification Method Purified
Applications Flow Cytometry