Please login to your online account to display your discounted pricing

Golden Guides


A student and teacher favorite.

Manufacturer: fisher science education  0307-240525

 View more versions of this product

Catalog No. S32093

  • / Each
 

Description & Specifications

Specifications

Format Paperback
Author Robert T. Mitchell, Herbert S. Zim
For Use With (Application) Information includes how to identify butterflies and moths
Product Type Butterflies and Moths
Category Biology