Please login to your online account to display your discounted pricing

Golden Guides


A student and teacher favorite.

Manufacturer: fisher science education  240534

 View more versions of this product

Catalog No. S51453

  • / Each
 

Description & Specifications

Specifications

Format Paperback
Product Type Birds
For Use With (Application) Full of colorful, magnificently illustrated guide gives information on Birds
Author Herbert S. Zim, Ira N. Gabrielson
Category Biology