Please login to your online account to display your discounted pricing

Golden Guides


A student and teacher favorite.

Manufacturer: fisher science education  24055

 View more versions of this product

Catalog No. S51316

  • / Each
 

Description & Specifications

Specifications

Product Type Insects
Author Clarence Cottam, Herbert S. Zim
Category Biology
For Use With (Application) Full of colorful, magnificently illustrated guide gives information on insect groups
Format Paperback