missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold wire, 99.9985% (metals basis), Alfa Aesar™, Premion™

Form: approx. 3.92g/m

Manufacturer:  Alfa Aesar 10966BQ

 View more versions of this product

Catalog No. AA10966BQ



Specifications

Specifications

Gold
7440-57-5
99.9985% (Metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
196.967
CHEBI:30050
Golden
5cm
Gold colored wire
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
Odorless
23985
Wire, 0.5mm (0.02 in.) dia.

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

EINECSNumber : 231-165-9

Documents