missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold wire, 2.0mm (0.08 in.) dia., Premion™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 14733BQ

 View more versions of this product

Catalog No. AA14733BQ



Specifications

Specifications

Gold wire
5cm
7440-57-5
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
Odorless
gold
23985
Wire
Premion™
≈60.6g/m
2.0mm (0.08 in.) dia.
(metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
[Au]
196.967
CHEBI:30050
99.999%

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents