missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold wire, 2.0mm (0.08 in.) dia., 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 14729BQ

 View more versions of this product

Catalog No. AA14729BQ



Specifications

Specifications

5cm
≈60.6g/m
7440-57-5
(metals basis)
MFCD00003436
Odorless
gold
23985
Wire
Gold wire
2.0mm (0.08 in.) dia.
99.95%
Au
PCHJSUWPFVWCPO-UHFFFAOYSA-N
[Au]
196.967
CHEBI:30050

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
CHEBI:30050
[Au]
Au
MFCD00003436
23985
gold
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents