missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold Wire, 1.0mm (0.04 in.) dia., Premion™, 99.999% (Metals basis), Alfa Aesar™

Form: Approx.15.15g/m

Manufacturer:  Alfa Aesar 14726BP

 View more versions of this product

Catalog No. AA14726BP



Specifications

Specifications

Gold
7440-57-5
99.999% (Metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
[Au]
196.967
CHEBI:30050
Golden
2cm
Gold colored wire
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
Odorless
gold
23985
Wire, 1.0mm (0.04 in.) dia.

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]
SDS Documents