missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold wire, 1.0mm (0.04 in.) dia., Premion™, 99.9985% (metals basis)

Manufacturer:  Alfa Aesar™ 10965BP

 View more versions of this product

Catalog No. AA10965BP



Specifications

Specifications

Gold wire
2cm
7440-57-5
Au
burnish
[Au]
Odorless
23985
196.97
99.9985%
≈15.15g/m
1.0mm (0.04 in.) dia.
(metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
196.967
CHEBI:30050
Wire
Premion™

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
burnish
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents