missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold wire, 1.0mm (0.04 in.) dia., 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 14731BW

 View more versions of this product

Catalog No. AA14731BW



Specifications

Specifications

Gold wire
50cm
7440-57-5
(metals basis)
MFCD00003436
Odorless
gold
23985
Wire
≈15.15g/m
1.0mm (0.04 in.) dia.
99.95%
Au
PCHJSUWPFVWCPO-UHFFFAOYSA-N
[Au]
196.967
CHEBI:30050

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
CHEBI:30050
[Au]
Au
MFCD00003436
23985
gold
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents