missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold wire, 0.813mm (0.032 in.) dia., 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 10197BS

 View more versions of this product

Catalog No. AA10197BS



Specifications

Specifications

Gold wire
10cm
7440-57-5
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
Odorless
23985
Wire
≈10.02g/m
0.813mm (0.032 in.) dia.
(metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
196.967
CHEBI:30050
99.9%

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents