missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold wire, 0.25mm (0.01 in.) dia., Premion™, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 10967BY

 View more versions of this product

Catalog No. AA10967BY



Specifications

Specifications

Gold wire
100cm
7440-57-5
Au
burnish
[Au]
Odorless
23985
Wire
Premion™
≈0.95g/m
0.25mm (0.01 in.) dia.
(metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
196.967
CHEBI:30050
99.998%

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
burnish
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents