missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold Wire, 0.25mm (0.01 in.) dia., 99.95% (Metals basis), Alfa Aesar™

Form: Approx.0.95g/m

Manufacturer:  Alfa Aesar 14724BY

 View more versions of this product

Catalog No. AA14724BY



Specifications

Specifications

Gold
7440-57-5
99.95% (Metals basis)
MFCD00003436
Odorless
gold
23985
Wire, 0.25mm (0.01 in.) dia.
100cm
Gold colored wire
Au
PCHJSUWPFVWCPO-UHFFFAOYSA-N
[Au]
196.967
CHEBI:30050
Golden

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
CHEBI:30050
[Au]
Au
MFCD00003436
23985
gold
SDS Documents