Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold wire, 0.0127mm (0.0005 in.) dia., 4.88m (16ft), 99.9% (metals basis)

$318.64 - $318.64

Chemical Identifiers

Chemical Name or Material Gold wire
CAS 7440-57-5
Name Note 0.0127mm (0.0005 in.) dia., 4.88m (16 ft.)
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14353KS

  SDS
alfa aesar™
14353KS*
1 each Each for $318.64
Description & Specifications

Specifications

Chemical Name or Material Gold wire
TSCA TSCA
Name Note 0.0127mm (0.0005 in.) dia., 4.88m (16 ft.)
Physical Form Wire
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00003436
CAS 7440-57-5
Odor Odorless
Weight ≈0.0024g/m
Quantity 1 each