missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold Thinfoil, 0.003mm (0.00012 in.) thick, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 44137FI

 View more versions of this product

Catalog No. AA44137FI



Specifications

Specifications

Gold Thinfoil
50 x 50mm
7440-57-5
Au
burnish
[Au]
Odorless
23985
Thin Foil, ≈0.04g/25x25mm,not certified pinhole free
≈0.04g/25x25mm
0.003mm (0.00012 in.) thick, not certified pinhole free
(metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
196.967
CHEBI:30050
99.9%

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
burnish
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

EINECSNumber : 231-165-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents