missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold slug, Premion™, 99.999% (metals basis), Alfa Aesar™

Form: approx. 3.35-3.60g/slug, approx. weight

Manufacturer:  Alfa Aesar 14174KJ

 View more versions of this product

Catalog No. AA14174KJ



Specifications

Specifications

Gold
7440-57-5
99.999% (Metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
196.967
CHEBI:30050
Golden
5 pc.
Gold colored slug
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
Odorless
23985
Slug, dia. X L: 6.35 X 6.35mm (0.25 X 0.25 in.)

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]
Safety and Handling

Safety and Handling

EINECSNumber : 231-165-9

Documents