missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold Slug, Dia. X L: 3.175 x 6.35mm (0.125 x 0.25 in.), Premion™, 99.99% (Metals basis), Alfa Aesar™

Form: Approx.0.97g/slug

Manufacturer:  Alfa Aesar 13394KJ

 View more versions of this product

Catalog No. AA13394KJ



Specifications

Specifications

Gold
7440-57-5
19.32g/cm3 at 20°C
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
Odorless
23985
Slug, dia. X L: 3.175 X 6.35mm (0.125 X 0.25 in.)
5 pc.
Gold colored slug
99.99% (Metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
196.967
CHEBI:30050
Golden

Chemical Identifiers

7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]
Documents