missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gold slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals
Form: Approx.0.97g/slug
Supplier: Thermo Scientific Chemicals 013394KM
Description
Gold slugs are used in thermal evaporation and electrodes (quartz oscillator, light-emitting diodes, and laser diodes).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Golden | |
7440-57-5 | |
19.32g/cm3 at 20°C | |
Au | |
25 Pc. | |
Soluble in hot sulfuric acid and aqua regia. Insoluble in water and acid. | |
[Au] | |
196.97 | |
CHEBI:30050 | |
Slug |
Gold | |
Gold colored slug | |
99.99% (Metals basis) | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
Odorless |
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
CHEBI:30050 | |
[Au] |
Safety and Handling
EINECSNumber : 231-165-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only