Please login to your online account to display your discounted pricing

Thermo Scientific™ Gold Seal™ Rite-On™ & Ultra Frost™ Frosted Microscope Slides


Label slides quickly and easily with Thermo Scientific™ Gold Seal™ Rite-On™ and  Ultra Frost™ Frosted Microscope slides, made from hard, colorless, noncorrosive glass.

Manufacturer: thermo scientific™  ER3017

 View more versions of this product

Catalog No. 12-518-100C

  • / Gross of 144 Each
  • / Case of 25 Gross
 

Description & Specifications

Specifications

Width (English) 1 in.
Edges Ground
Corner Style 90°
Dimensions (L x W) 3 x 1 in. (75 x 25mm)
Coating Double Sandblast Frosted End, Both Sides
Length (English) 3 in.
Packaging Quantity Packed 72 slides per box, 50 boxes per case
Description Rite-On Frosted Slide
Length (Metric) 75mm
Thickness (Metric) 1mm
Width (Metric) 25mm

Label slides quickly and easily with Thermo Scientific™ Gold Seal™ Rite-On™ and  Ultra Frost™ Frosted Microscope slides, made from hard, colorless, noncorrosive glass. Choose from sandblast frosted end or white epoxy frosted end for easy marking. Each tab is impervious to virtually all common laboratory chemicals and reagents.

Accessories

Accessories for Catalog Number: 12518100C

Item Mfr. No. Item Description Price Per Unit Quantity  
03-448-6
Fisherbrand™ Colored ABS 25 Place Slide Boxes
fisherbrand™
HS15989MF
Store and transport 25 slides safely with durable ABS cases. Fisherbrand™ Colored ABS 25 Place Slide Boxes keep standard microscope slides in numbered, grooved slots. Provides safe, dust-free storage. Each for $6.36
Case of 72 Each for $457.92