Please login to your online account to display your discounted pricing
Thermo Scientific™ Gold Seal™ Beveled Microslides
Produce margin-free smears on Thermo Scientific™ Gold Seal™ Beveled Microslides, whose diamond-ground beveled edges allow for easy handling.
$90.59 - $3,287.03
Specifications
Length (Metric) 75mm
Width (Metric) 25mm
View More Specs
Products
Catalog Number Mfr. No. Frosted Size Price Quantity    
12-518-110A
  Accessories
thermo scientific™
ER3057
No Gross of 144 Each for $90.59
Case of 25 Gross for $2,264.75
12-518-110B
  Accessories
thermo scientific™
ER3058
Yes 75 x 25mm Gross of 144 Each for $123.38
Case of 25 Gross for $3,084.50
12-518-100D
thermo scientific™
ER3061002
No Gross of 144 Each for $98.68
Case of 25 Gross for $2,467.00
12-518-104B
  Accessories
thermo scientific™
ER3062
Yes 3 x 1 in. Case of 3600 Each for $3,287.03
Description & Specifications
Specifications
Length (Metric) 75mm
Width (Metric) 25mm
Mount specimens on Thermo Scientific™ Gold Seal™ Plain Microscope slides, made from the finest white sheet glass, where each slide is selected for uniform quality. Gold Seal Plain Microscope slides are made from hard, colorless, noncorrosive glass. All slides feature ground and polished edges that allow for easy handling.
All finished slides are sealed with a desiccant in an air tight, lint-free, hermetically sealed Moistur-Barrier™ package, keeping slides dry during shipping and storage.
Slides work well in hematology, microbiology, and urinalysis as well as other laboratory and research applications.
Features:
  • Slides are available in plain glass or frosted on both sides at one end
  • Pure white glass provides better light transmission
  • Diamond-ground edges offer smoothness and uniformity
  • Clipped-corner slides are 1mm thick
  • Optimum feathering is easy on fingers
  • Each case features an inner barrier-bag sealed with desiccant packs to protect from moisture contamination
  • Slide boxes convert into convenient slide storage files
  • Reduced iron content for low self-fluorescence
  • Manufactured from pure, water-white glass
  • Precleaned for immediate use
  • Gold Seal is a trademark for Becton, Dickinson and Company.