Thermo Scientific™ Gold Seal™ Rite-On™ and Ultra Frost™ Frosted Microscope Slides
Label slides quickly and easily with Thermo Scientific™ Gold Seal™ Rite-On™ and  Ultra Frost™ Frosted Microscope slides, made from hard, colorless, noncorrosive glass.
Manufacturer:  Thermo Scientific™ ER3058
 View more versions of this product
Catalog No. 12-518-110B
Specifications
Description
Label slides quickly and easily with Thermo Scientific™ Gold Seal™ Rite-On™ and  Ultra Frost™ Frosted Microscope slides, made from hard, colorless, noncorrosive glass. Choose from sandblast frosted end or white epoxy frosted end for easy marking. Each tab is impervious to virtually all common laboratory chemicals and reagents.
Description & Specifications
Specifications
Rite-On Frosted Beveled Slide with Clipped Corners
Beveled
75mm
25mm
45° Clipped
Packed 72 slides per box, 50 boxes per case
1mm
3 in.
1 in.
Double Sandblast Frosted End, Both Sides
3 x 1 in. (75 x 25mm)
Accessories