Please login to your online account to display your discounted pricing

Alfa Aesar™ Gold rod, 6.35mm (0.25 in.) dia., Premion™, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Gold rod
CAS 7440-57-5
Name Note 6.35mm (0.25 in.) dia.
Purity Grade Premion™
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12555HF

  SDS
alfa aesar™
12555HF
10mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gold rod
TSCA TSCA
Name Note 6.35mm (0.25 in.) dia.
Purity Grade Premion™
Physical Form Rod
Assay 99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00003436
CAS 7440-57-5
Odor Odorless
Weight ≈6.1g/10mm
Quantity 10mm