Please login to your online account to display your discounted pricing

Gold powder, spherical, APS 0.8-1.5 micron, 99.96+% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Gold powder
CAS 7440-57-5
Name Note spherical, APS 0.8 to 1.5μm
MDL Number MFCD00003436
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3981706

  SDS
alfa aesar
3981706
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Name Note spherical, APS 0.8 to 1.5μm
Assay Percent Notes (metals basis)
Chemical Name or Material Gold powder
Assay ≥99.96%
MDL Number MFCD00003436
Surface Area 0.15 to 0.45m2/g
TSCA TSCA
CAS 7440-57-5